Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTGTCCATTTCATTGTTTATTTCA[A/G]TACCATGTATAGTATCTGAATTGTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 608672 MIM: 610643 | ||||||||||||||||||||
Literature Links: |
DZIP3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DZIP3 - DAZ interacting zinc finger protein 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014648.3 | Intron | NP_055463.1 | ||||
XM_005247914.3 | Intron | XP_005247971.1 | ||||
XM_005247915.3 | Intron | XP_005247972.1 | ||||
XM_005247916.3 | Intron | XP_005247973.1 | ||||
XM_005247917.3 | Intron | XP_005247974.1 | ||||
XM_005247918.3 | Intron | XP_005247975.1 | ||||
XM_005247919.3 | Intron | XP_005247976.1 | ||||
XM_017007540.1 | Intron | XP_016863029.1 |
KIAA1524 - KIAA1524 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |