Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGGTGTTAGTGTATCCTGAGTTAC[A/G]GGGGCCTGGCAGAAGAAAGACGACA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603397 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF282 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ZNF282 - zinc finger protein 282 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001303481.1 | Intron | NP_001290410.1 | ||||
NM_003575.3 | Intron | NP_003566.1 | ||||
XM_006716151.3 | Intron | XP_006716214.1 |
Set Membership: |
HapMap |