Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTCAAACTTGGATTTTCTGACTCT[A/G]TAAATTCAATTCTATTTCATATGTA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606245 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SUZ12 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
SUZ12 - SUZ12 polycomb repressive complex 2 subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001321207.1 | Intron | NP_001308136.1 | ||||
NM_015355.3 | Intron | NP_056170.2 | ||||
XM_006721794.3 | Intron | XP_006721857.1 | ||||
XM_017024409.1 | Intron | XP_016879898.1 |
Set Membership: |
HapMap |