Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTTATATGAACAACAAGACTGCAC[G/T]TTCCCATCCAGCCTTAGGTGGCACA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602329 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SEL1L PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
SEL1L - SEL1L ERAD E3 ligase adaptor subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001244984.1 | 4604 | Intron | NP_001231913.1 | |||
NM_005065.5 | 4604 | UTR 3 | NP_005056.3 | |||
XM_005267988.2 | 4604 | UTR 3 | XP_005268045.1 | |||
XM_005267989.3 | 4604 | UTR 3 | XP_005268046.1 |