Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCAGATGAGGGTAGAATCAGATTG[G/T]TTAAATGTAGATGAGCCATGAGCTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
C16orf70 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
C16orf70 - chromosome 16 open reading frame 70 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001320540.1 | Intron | NP_001307469.1 | ||||
NM_001320541.1 | Intron | NP_001307470.1 | ||||
NM_001320542.1 | Intron | NP_001307471.1 | ||||
NM_001320543.1 | Intron | NP_001307472.1 | ||||
NM_025187.4 | Intron | NP_079463.2 | ||||
XM_006721284.3 | Intron | XP_006721347.1 | ||||
XM_011523350.2 | Intron | XP_011521652.1 | ||||
XM_017023729.1 | Intron | XP_016879218.1 | ||||
XM_017023730.1 | Intron | XP_016879219.1 | ||||
XM_017023731.1 | Intron | XP_016879220.1 |