Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGACTCCACCACCATAAAGGCGAA[A/C]CCGCACTGACGGAGAGGAGCCAGCG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
ZNF561 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ZNF561 - zinc finger protein 561 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152289.2 | 11 | UTR 5 | NP_689502.2 | |||
XM_005260150.2 | 11 | UTR 5 | XP_005260207.1 | |||
XM_005260151.2 | 11 | UTR 5 | XP_005260208.1 | |||
XM_006722938.2 | 11 | UTR 5 | XP_006723001.1 | |||
XM_011528433.2 | 11 | UTR 5 | XP_011526735.1 | |||
XM_011528434.2 | 11 | UTR 5 | XP_011526736.1 | |||
XM_011528436.2 | 11 | UTR 5 | XP_011526738.1 | |||
XM_011528437.2 | 11 | UTR 5 | XP_011526739.1 | |||
XM_017027478.1 | 11 | UTR 5 | XP_016882967.1 | |||
XM_017027479.1 | 11 | UTR 5 | XP_016882968.1 | |||
XM_017027480.1 | 11 | UTR 5 | XP_016882969.1 | |||
XM_017027481.1 | 11 | Intron | XP_016882970.1 | |||
XM_017027482.1 | 11 | UTR 5 | XP_016882971.1 |
ZNF561-AS1 - ZNF561 antisense RNA 1 (head to head) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |