Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTGTGCTGTGCACCATGGTCCACG[A/G]ACTGGCATCTTCCCCACTCGCGGTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 147840 MIM: 614088 MIM: 601852 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ICAM1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ICAM1 - intercellular adhesion molecule 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ICAM4 - intercellular adhesion molecule 4 (Landsteiner-Wiener blood group) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ICAM5 - intercellular adhesion molecule 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003259.3 | Intron | NP_003250.3 | ||||
XM_011528229.1 | Intron | XP_011526531.1 | ||||
XM_017027185.1 | Intron | XP_016882674.1 |