Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGTTTGACCCTAGCAAGCCTTTCT[C/T]TCTCACTTCCCGAGGTGCCTTGTTT
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 133520 | |||||||||||||||||||||||
Literature Links: |
ERCC4 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU)
|
|||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
ERCC4 - ERCC excision repair 4, endonuclease catalytic subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005236.2 | 1449 | Missense Mutation | TCT,TTT | S,F 768 | NP_005227.1 | |
XM_011522424.2 | 1449 | Missense Mutation | TCT,TTT | S,F 814 | XP_011520726.1 | |
XM_011522427.1 | 1449 | Missense Mutation | TCT,TTT | S,F 318 | XP_011520729.1 | |
XM_017023043.1 | 1449 | Missense Mutation | TCT,TTT | S,F 505 | XP_016878532.1 |
Set Membership: |
HapMap |