Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 141251 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
HMOX2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
HMOX2 - heme oxygenase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001127204.1 | Intron | NP_001120676.1 | ||||
NM_001127205.1 | Intron | NP_001120677.1 | ||||
NM_001127206.2 | Intron | NP_001120678.1 | ||||
NM_001286267.1 | Intron | NP_001273196.1 | ||||
NM_001286268.1 | Intron | NP_001273197.1 | ||||
NM_001286269.1 | Intron | NP_001273198.1 | ||||
NM_001286270.1 | Intron | NP_001273199.1 | ||||
NM_001286271.1 | Intron | NP_001273200.1 | ||||
NM_002134.3 | Intron | NP_002125.3 | ||||
XM_011522473.1 | Intron | XP_011520775.1 | ||||
XM_017023196.1 | Intron | XP_016878685.1 | ||||
XM_017023197.1 | Intron | XP_016878686.1 |
NMRAL1 - NmrA-like family domain containing 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |