Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCCAGGTAGGGCGCCAACAGCATC[A/C]AGCTGTGCGCGTGGGCTCTGCGCAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 194521 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF33A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZNF33A - zinc finger protein 33A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278170.1 | Intron | NP_001265099.1 | ||||
NM_001278171.1 | Intron | NP_001265100.1 | ||||
NM_001278173.1 | Intron | NP_001265102.1 | ||||
NM_001278174.1 | Intron | NP_001265103.1 | ||||
NM_001278175.1 | Intron | NP_001265104.1 | ||||
NM_001278176.1 | Intron | NP_001265105.1 | ||||
NM_001278177.1 | Intron | NP_001265106.1 | ||||
NM_001278178.1 | Intron | NP_001265107.1 | ||||
NM_001278179.1 | Intron | NP_001265108.1 | ||||
NM_001324175.1 | Intron | NP_001311104.1 | ||||
NM_001324176.1 | Intron | NP_001311105.1 | ||||
NM_001324177.1 | Intron | NP_001311106.1 | ||||
NM_001324178.1 | Intron | NP_001311107.1 | ||||
NM_006954.1 | Intron | NP_008885.1 | ||||
NM_006974.2 | Intron | NP_008905.1 | ||||
XM_011519650.2 | Intron | XP_011517952.1 | ||||
XM_011519651.2 | Intron | XP_011517953.1 | ||||
XM_011519653.2 | Intron | XP_011517955.1 | ||||
XM_011519654.2 | Intron | XP_011517956.1 | ||||
XM_017016617.1 | Intron | XP_016872106.1 |