Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATGACAGGTAGTAATCTGAGAACA[A/G]GCATAGATAATTGGTTCATTTTCTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608668 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZMYND11 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ZMYND11 - zinc finger MYND-type containing 11 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001202464.1 | Intron | NP_001189393.1 | ||||
NM_001202465.1 | Intron | NP_001189394.1 | ||||
NM_001202466.1 | Intron | NP_001189395.1 | ||||
NM_001202467.1 | Intron | NP_001189396.1 | ||||
NM_001202468.1 | Intron | NP_001189397.1 | ||||
NM_006624.5 | Intron | NP_006615.2 | ||||
NM_212479.3 | Intron | NP_997644.2 | ||||
XM_005252359.4 | Intron | XP_005252416.1 | ||||
XM_005252361.3 | Intron | XP_005252418.1 | ||||
XM_005252362.2 | Intron | XP_005252419.1 | ||||
XM_006717376.2 | Intron | XP_006717439.1 | ||||
XM_017015587.1 | Intron | XP_016871076.1 | ||||
XM_017015588.1 | Intron | XP_016871077.1 | ||||
XM_017015589.1 | Intron | XP_016871078.1 | ||||
XM_017015590.1 | Intron | XP_016871079.1 | ||||
XM_017015591.1 | Intron | XP_016871080.1 | ||||
XM_017015592.1 | Intron | XP_016871081.1 | ||||
XM_017015593.1 | Intron | XP_016871082.1 | ||||
XM_017015594.1 | Intron | XP_016871083.1 |
Set Membership: |
HapMap |