Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTTTGAAGTCCAAGTGGCTCCAGA[A/G]GAAACAGTTGAGGTAAGATAATGGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
OR13A1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
OR13A1 - olfactory receptor family 13 subfamily A member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001004297.2 | 5579 | UTR 3 | NP_001004297.2 | |||
XM_011540147.2 | 5579 | UTR 3 | XP_011538449.1 | |||
XM_017016632.1 | 5579 | UTR 3 | XP_016872121.1 | |||
XM_017016633.1 | 5579 | UTR 3 | XP_016872122.1 | |||
XM_017016634.1 | 5579 | UTR 3 | XP_016872123.1 |
Set Membership: |
HapMap |