Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602217 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SDCBP PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
SDCBP - syndecan binding protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001007067.1 | Intron | NP_001007068.1 | ||||
NM_001007068.1 | Intron | NP_001007069.1 | ||||
NM_001007069.1 | Intron | NP_001007070.1 | ||||
NM_001007070.1 | Intron | NP_001007071.1 | ||||
NM_005625.3 | Intron | NP_005616.2 | ||||
XM_011517571.2 | Intron | XP_011515873.2 | ||||
XM_017013727.1 | Intron | XP_016869216.1 | ||||
XM_017013728.1 | Intron | XP_016869217.1 |
Set Membership: |
HapMap |