Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTTTCCTTTTCTCAAAACAGTAGCA[A/G]ATTTCTGAAATATTAGAAGTTCCAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603711 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CYP7B1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CYP7B1 - cytochrome P450 family 7 subfamily B member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001324112.1 | 3002 | Intron | NP_001311041.1 | |||
NM_004820.4 | 3002 | UTR 3 | NP_004811.1 | |||
XM_017014002.1 | 3002 | UTR 3 | XP_016869491.1 |
Set Membership: |
HapMap |