Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCGCCATGGATTGTGGCCAAATGA[C/G]GTTGAGAGACGATGTTTTCACCTGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609084 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FBXL18 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
FBXL18 - F-box and leucine rich repeat protein 18 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001321213.1 | Intron | NP_001308142.1 | ||||
NM_024963.5 | Intron | NP_079239.3 | ||||
XM_011515531.2 | Intron | XP_011513833.1 | ||||
XM_011515532.2 | Intron | XP_011513834.1 |