Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTATCCATAATAATAATGACGATGG[C/T]GATAATTTAGAGCTGTAGTATGTTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 176385 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PAPPA PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PAPPA - pappalysin 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002581.4 | Intron | NP_002572.2 | ||||
XM_006717129.2 | Intron | XP_006717192.1 | ||||
XM_017014784.1 | Intron | XP_016870273.1 |