Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGGACCGCGCCGGGCGCAAGCTGA[G/T]CCTCTTGCTGTGCTCCGTGCCCTTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605245 | ||||||||||||||||||||
Literature Links: |
GARNL3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GARNL3 - GTPase activating Rap/RanGAP domain like 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLC2A8 - solute carrier family 2 member 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001271711.1 | 389 | Missense Mutation | AGC,ATC | S,I 99 | NP_001258640.1 | |
NM_001271712.1 | 389 | Intron | NP_001258641.1 | |||
NM_014580.4 | 389 | Missense Mutation | AGC,ATC | S,I 99 | NP_055395.2 | |
XM_006717084.3 | 389 | Missense Mutation | AGC,ATC | S,I 99 | XP_006717147.1 | |
XM_011518602.2 | 389 | Missense Mutation | AGC,ATC | S,I 99 | XP_011516904.1 | |
XM_011518603.2 | 389 | Intron | XP_011516905.1 | |||
XM_011518604.2 | 389 | Intron | XP_011516906.1 | |||
XM_017014653.1 | 389 | Intron | XP_016870142.1 |