Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTCCCCTCCCCCTCCAGCCCTGGC[A/G]TCTCCCCTCTTCCTCTCCCCAGGAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600547 MIM: 608360 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
KYAT1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KYAT1 - kynurenine aminotransferase 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LRRC8A - leucine rich repeat containing 8 family member A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001127244.1 | Intron | NP_001120716.1 | ||||
NM_001127245.1 | Intron | NP_001120717.1 | ||||
NM_019594.3 | Intron | NP_062540.2 | ||||
XM_005252095.2 | Intron | XP_005252152.1 | ||||
XM_005252096.3 | Intron | XP_005252153.1 | ||||
XM_006717186.2 | Intron | XP_006717249.1 | ||||
XM_011518863.1 | Intron | XP_011517165.1 | ||||
XM_011518864.2 | Intron | XP_011517166.1 | ||||
XM_011518865.2 | Intron | XP_011517167.1 |