Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGGGCTTCGTGGATGCCCTGCACC[C/G]GGTCCGGCAGGTACGGGCGCAGCCG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603536 | ||||||||||||||||||||
Literature Links: |
FUBP3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FUBP3 - far upstream element binding protein 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003934.1 | 188 | Missense Mutation | CCG,CGG | P,R 25 | NP_003925.1 | |
XM_005272232.1 | 188 | Missense Mutation | CCG,CGG | P,R 25 | XP_005272289.1 | |
XM_006717313.2 | 188 | UTR 5 | XP_006717376.1 | |||
XM_011519172.2 | 188 | Missense Mutation | CCG,CGG | P,R 25 | XP_011517474.1 | |
XM_011519173.2 | 188 | Intron | XP_011517475.1 | |||
XM_011519174.2 | 188 | UTR 5 | XP_011517476.1 | |||
XM_017015269.1 | 188 | UTR 5 | XP_016870758.1 | |||
XM_017015270.1 | 188 | UTR 5 | XP_016870759.1 | |||
XM_017015271.1 | 188 | UTR 5 | XP_016870760.1 | |||
XM_017015272.1 | 188 | UTR 5 | XP_016870761.1 | |||
XM_017015273.1 | 188 | UTR 5 | XP_016870762.1 |
LOC100272217 - uncharacterized LOC100272217 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |