Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAATCACAAGACATAAAAGTAAAA[C/T]GTGATAAATCGGCTGTCATCTAGTT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604689 MIM: 609181 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
AKAP3 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
AKAP3 - A-kinase anchoring protein 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278309.1 | Intron | NP_001265238.1 | ||||
NM_006422.3 | Intron | NP_006413.3 | ||||
XM_005253664.2 | Intron | XP_005253721.1 | ||||
XM_011520908.1 | Intron | XP_011519210.1 | ||||
XM_011520909.1 | Intron | XP_011519211.1 | ||||
XM_011520910.2 | Intron | XP_011519212.1 |
DYRK4 - dual specificity tyrosine phosphorylation regulated kinase 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |