Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCTCCATTCCGATATCTGGGCTGC[A/C]CTGATCTACCACTCTCCTGGTTTCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616082 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
C12orf4 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
C12orf4 - chromosome 12 open reading frame 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001304811.1 | 3147 | UTR 3 | NP_001291740.1 | |||
NM_020374.3 | 3147 | UTR 3 | NP_065107.1 | |||
XM_005253716.1 | 3147 | UTR 3 | XP_005253773.1 | |||
XM_006718992.3 | 3147 | Intron | XP_006719055.1 |
Set Membership: |
HapMap |