Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GATCACTTCTGCTGAGTATCTCTTC[C/G]TCTTACTTCTGCCACTGTCCCATGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609046 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
BTBD17 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
BTBD17 - BTB domain containing 17 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001080466.1 | Intron | NP_001073935.1 | ||||
XM_011524791.2 | Intron | XP_011523093.1 | ||||
XM_017024622.1 | Intron | XP_016880111.1 |
GPR142 - G protein-coupled receptor 142 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
KIF19 - kinesin family member 19 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |