Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCAAGACAGTCTGTGCCGCCAAGC[C/T]GGCCAGAGCTTGCAGAGCTCCCCTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC105274304 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
LOC105274304 - uncharacterized LOC105274304 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MXRA7 - matrix remodeling associated 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001008528.2 | Intron | NP_001008528.1 | ||||
NM_001008529.2 | Intron | NP_001008529.1 | ||||
NM_198530.3 | Intron | NP_940932.2 | ||||
XM_005257382.2 | Intron | XP_005257439.1 | ||||
XM_005257383.2 | Intron | XP_005257440.1 | ||||
XM_005257384.2 | Intron | XP_005257441.1 | ||||
XM_011524830.1 | Intron | XP_011523132.1 |