Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 182115 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CYTH1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CYTH1 - cytohesin 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001292018.1 | Intron | NP_001278947.1 | ||||
NM_001292019.1 | Intron | NP_001278948.1 | ||||
NM_004762.3 | Intron | NP_004753.1 | ||||
NM_017456.3 | Intron | NP_059430.2 | ||||
XM_011525475.2 | Intron | XP_011523777.1 | ||||
XM_011525476.2 | Intron | XP_011523778.1 | ||||
XM_011525477.1 | Intron | XP_011523779.1 | ||||
XM_011525478.1 | Intron | XP_011523780.1 | ||||
XM_017025336.1 | Intron | XP_016880825.1 | ||||
XM_017025337.1 | Intron | XP_016880826.1 | ||||
XM_017025338.1 | Intron | XP_016880827.1 |
Set Membership: |
HapMap |