Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCCTTAATGTATACAGTCCATACT[C/T]ATAAGGGAAACAAGAATTGTCTTCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
26 submissions
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613292 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DENND1B PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
DENND1B - DENN domain containing 1B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001195215.1 | 4120 | UTR 3 | NP_001182144.1 | |||
NM_001195216.1 | 4120 | Intron | NP_001182145.1 | |||
NM_001300858.1 | 4120 | Intron | NP_001287787.1 | |||
NM_144977.4 | 4120 | Intron | NP_659414.2 | |||
XM_005244931.2 | 4120 | UTR 3 | XP_005244988.1 | |||
XM_006711192.2 | 4120 | UTR 3 | XP_006711255.1 | |||
XM_006711193.3 | 4120 | UTR 3 | XP_006711256.1 | |||
XM_006711194.3 | 4120 | UTR 3 | XP_006711257.1 | |||
XM_011509246.2 | 4120 | UTR 3 | XP_011507548.1 | |||
XM_011509247.1 | 4120 | UTR 3 | XP_011507549.1 | |||
XM_011509248.2 | 4120 | UTR 3 | XP_011507550.1 | |||
XM_011509249.2 | 4120 | UTR 3 | XP_011507551.1 | |||
XM_011509251.2 | 4120 | Intron | XP_011507553.1 | |||
XM_017000470.1 | 4120 | UTR 3 | XP_016855959.1 | |||
XM_017000471.1 | 4120 | UTR 3 | XP_016855960.1 | |||
XM_017000472.1 | 4120 | UTR 3 | XP_016855961.1 |