Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTTATGGACTTTGGGGTCAGTTCA[C/G]ATTCTGCCTCTTCCACTCAGTAACT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
27 submissions
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 169190 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CDK18 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CDK18 - cyclin dependent kinase 18 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002596.3 | Intron | NP_002587.2 | ||||
NM_212502.2 | Intron | NP_997667.1 | ||||
NM_212503.2 | Intron | NP_997668.1 | ||||
XM_011509602.2 | Intron | XP_011507904.1 | ||||
XM_017001423.1 | Intron | XP_016856912.1 |
Set Membership: |
HapMap |