Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGTCTCAAGGCAGGAGAAGAAGAA[C/T]ATGTGCTGTTTTCCTGGGAGTGGGG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
31 submissions
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614503 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
KLHDC8A PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese)
|
||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
KLHDC8A - kelch domain containing 8A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001271863.1 | 2675 | UTR 3 | NP_001258792.1 | |||
NM_001271864.1 | 2675 | UTR 3 | NP_001258793.1 | |||
NM_001271865.1 | 2675 | UTR 3 | NP_001258794.1 | |||
NM_018203.2 | 2675 | UTR 3 | NP_060673.1 |