Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAATAAACTAAGAGTTCACAGAACA[A/G]CTAAGAGAACTTAAACCCAAGCCTA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608813 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DERL1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
DERL1 - derlin 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001134671.2 | Intron | NP_001128143.1 | ||||
NM_024295.5 | Intron | NP_077271.1 | ||||
XM_006716657.1 | Intron | XP_006716720.1 | ||||
XM_011517299.1 | Intron | XP_011515601.1 |
Set Membership: |
HapMap |