Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGGCATCTGAGTAACTTCGCTGCT[A/C]CAACTGTGGACTTGGTCCCTTGCTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
54 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611350 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
INTS7 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
INTS7 - integrator complex subunit 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199809.1 | Intron | NP_001186738.1 | ||||
NM_001199811.1 | Intron | NP_001186740.1 | ||||
NM_001199812.1 | Intron | NP_001186741.1 | ||||
NM_015434.3 | Intron | NP_056249.1 | ||||
XM_011509396.2 | Intron | XP_011507698.1 | ||||
XM_017000962.1 | Intron | XP_016856451.1 |