Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGAAAAAAATTATCACAGAATTTCA[C/T]AGACATATGCGAAACCCAAGGTAAT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616334 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TMEM100 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
TMEM100 - transmembrane protein 100 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001099640.1 | 3011 | UTR 3 | NP_001093110.1 | |||
NM_018286.2 | 3011 | UTR 3 | NP_060756.2 | |||
XM_017024814.1 | 3011 | UTR 3 | XP_016880303.1 | |||
XM_017024815.1 | 3011 | UTR 3 | XP_016880304.1 | |||
XM_017024816.1 | 3011 | UTR 3 | XP_016880305.1 |
Set Membership: |
HapMap |