Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCATTCCAAAATAGTATAGTTTGAC[C/G]TATCAGAGACCACAGCCATGGCCCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614693 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ATMIN PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ATMIN - ATM interactor | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
C16orf46 - chromosome 16 open reading frame 46 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001100873.1 | 2166 | UTR 3 | NP_001094343.1 | |||
NM_152337.2 | 2166 | Intron | NP_689550.2 | |||
XM_005255792.2 | 2166 | UTR 3 | XP_005255849.1 | |||
XM_011522848.2 | 2166 | UTR 3 | XP_011521150.2 | |||
XM_011522849.2 | 2166 | UTR 3 | XP_011521151.2 | |||
XM_017022916.1 | 2166 | UTR 3 | XP_016878405.1 | |||
XM_017022917.1 | 2166 | UTR 3 | XP_016878406.1 |