Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATATGTGGGCAAAACCTCTTCTCTA[C/T]TAAAATAAAATACCAATGAAAATAC
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608870 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LRIG3 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
LRIG3 - leucine rich repeats and immunoglobulin like domains 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001136051.2 | 6392 | Intron | NP_001129523.1 | |||
NM_153377.4 | 6392 | Intron | NP_700356.2 | |||
XM_011537893.2 | 6392 | UTR 3 | XP_011536195.1 | |||
XM_017018790.1 | 6392 | Intron | XP_016874279.1 |
Set Membership: |
HapMap |