Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCTGCTGATGGTCTCTCCCGCCGC[C/T]CTAGCCTCCTTCACCAATCCATTGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 615856 | ||||||||||||||||||||
Literature Links: |
TMTC2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TMTC2 - transmembrane and tetratricopeptide repeat containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001320321.1 | Intron | NP_001307250.1 | ||||
NM_001320322.1 | Intron | NP_001307251.1 | ||||
NM_152588.2 | Intron | NP_689801.1 | ||||
XM_017018884.1 | Intron | XP_016874373.1 |