Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCACAAATGGTAGTAATTATCATCA[A/C]CATCATCATCTTCTTAATCACCATC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608561 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
STAB2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
STAB2 - stabilin 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_017564.9 | Intron | NP_060034.9 | ||||
XM_011538537.2 | Intron | XP_011536839.1 | ||||
XM_011538538.2 | Intron | XP_011536840.1 | ||||
XM_011538539.2 | Intron | XP_011536841.1 | ||||
XM_011538541.2 | Intron | XP_011536843.1 | ||||
XM_011538542.2 | Intron | XP_011536844.1 | ||||
XM_017019585.1 | Intron | XP_016875074.1 |