Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TATTATTTGCATTTTGCAAAATGAA[A/G]AAATTGAGGTTTAGAGATACCCAAG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 176876 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PTPN11 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese)
|
||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
PTPN11 - protein tyrosine phosphatase, non-receptor type 11 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_006719526.2 | Intron | XP_006719589.1 | ||||
XM_011538613.2 | Intron | XP_011536915.1 | ||||
XM_017019722.1 | Intron | XP_016875211.1 |
Set Membership: |
HapMap |