Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATACAGGTTTGGAGTCACACCGTA[G/T]AGCTGAAAGCTTGGCTTTTATTTAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 609666 | ||||||||||||||||||||
Literature Links: |
IQCD PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
IQCD - IQ motif containing D | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TPCN1 - two pore segment channel 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001143819.2 | Intron | NP_001137291.1 | ||||
NM_001301214.1 | Intron | NP_001288143.1 | ||||
NM_017901.5 | Intron | NP_060371.2 | ||||
XM_005253906.4 | Intron | XP_005253963.1 | ||||
XM_011538490.2 | Intron | XP_011536792.1 | ||||
XM_011538492.2 | Intron | XP_011536794.1 | ||||
XM_011538493.2 | Intron | XP_011536795.1 | ||||
XM_017019480.1 | Intron | XP_016874969.1 | ||||
XM_017019481.1 | Intron | XP_016874970.1 | ||||
XM_017019482.1 | Intron | XP_016874971.1 |