Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATCACTAAAACAGGTAAACTTGCAG[C/G]TGCTCTCGCGAATCAGCTGGTAAGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604482 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
RNU4-1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RNU4-1 - RNA, U4 small nuclear 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RNU4-2 - RNA, U4 small nuclear 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SIRT4 - sirtuin 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_012240.2 | Intron | NP_036372.1 | ||||
XM_005253865.4 | Intron | XP_005253922.1 | ||||
XM_006719308.2 | Intron | XP_006719371.1 | ||||
XM_006719309.3 | Intron | XP_006719372.1 |