Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGAGATGTCTTACATACCTGTCTA[A/G]AGAACCTGTCTAGAGACCAACCATT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
LOC105370027 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LOC105370027 - uncharacterized LOC105370027 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMEM233 - transmembrane protein 233 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001136534.1 | Intron | NP_001130006.1 | ||||
XM_005253880.3 | Intron | XP_005253937.1 | ||||
XM_005253881.3 | Intron | XP_005253938.1 | ||||
XM_011538328.2 | Intron | XP_011536630.1 | ||||
XM_011538329.2 | Intron | XP_011536631.1 | ||||
XM_011538330.2 | Intron | XP_011536632.1 | ||||
XM_011538331.2 | Intron | XP_011536633.1 |