Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615806 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC15A4 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
SLC15A4 - solute carrier family 15 member 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_145648.3 | 2444 | UTR 3 | NP_663623.1 | |||
XM_011537895.1 | 2444 | UTR 3 | XP_011536197.1 | |||
XM_017018791.1 | 2444 | Intron | XP_016874280.1 | |||
XM_017018792.1 | 2444 | Intron | XP_016874281.1 | |||
XM_017018793.1 | 2444 | Intron | XP_016874282.1 |
Set Membership: |
HapMap |