Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTCGGCTCACCGCAAACTCTGCCT[C/T]CCGGGTTCAAGCAATTCTCCTGCCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC107985657 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC107985657 - uncharacterized LOC107985657 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TCEANC - transcription elongation factor A N-terminal and central domain containing | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001297563.1 | 272 | UTR 5 | NP_001284492.1 | |||
NM_001297564.1 | 272 | Intron | NP_001284493.1 | |||
NM_152634.3 | 272 | Silent Mutation | CTC,CTT | L,L 10 | NP_689847.2 | |
XM_017029316.1 | 272 | Silent Mutation | CTC,CTT | L,L 10 | XP_016884805.1 | |
XM_017029317.1 | 272 | Intron | XP_016884806.1 | |||
XM_017029318.1 | 272 | Intron | XP_016884807.1 | |||
XM_017029319.1 | 272 | Intron | XP_016884808.1 |