Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAATTGCTTCTGGAAAGCTTGTCA[A/G]TGAGTCCAGCATCCAGATTCTTTAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300051 MIM: 300170 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GPM6B PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
GPM6B - glycoprotein M6B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001001994.2 | 3583 | UTR 3 | NP_001001994.1 | |||
NM_001001995.2 | 3583 | UTR 3 | NP_001001995.1 | |||
NM_001001996.2 | 3583 | UTR 3 | NP_001001996.1 | |||
NM_001318729.1 | 3583 | UTR 3 | NP_001305658.1 | |||
NM_005278.4 | 3583 | UTR 3 | NP_005269.1 | |||
XM_005274489.4 | 3583 | UTR 3 | XP_005274546.1 | |||
XM_011545497.2 | 3583 | UTR 3 | XP_011543799.1 | |||
XM_017029432.1 | 3583 | UTR 3 | XP_016884921.1 | |||
XM_017029433.1 | 3583 | UTR 3 | XP_016884922.1 | |||
XM_017029434.1 | 3583 | UTR 3 | XP_016884923.1 |
OFD1 - OFD1, centriole and centriolar satellite protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003611.2 | 3583 | Intron | NP_003602.1 |
Set Membership: |
HapMap |