Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCTTTCCTTTCCTTAAGATCAATG[C/T]CTCATCTGAGCTGGAAAAGGAGCAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604197 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MTHFS PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MTHFS - 5,10-methenyltetrahydrofolate synthetase (5-formyltetrahydrofolate cyclo-ligase) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ST20 - suppressor of tumorigenicity 20 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001100879.1 | Intron | NP_001094349.1 | ||||
NM_001100880.2 | Intron | NP_001094350.1 | ||||
NM_001199757.1 | Intron | NP_001186686.1 |
ST20-MTHFS - ST20-MTHFS readthrough | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199760.1 | Intron | NP_001186689.1 |