Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTATTTTAATTCATTGTTTGAATGG[A/T]TATGTCTCTGACTATTATGGTATCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 614361 | ||||||||||||||||||||
Literature Links: |
ACSM5 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ACSM5 - acyl-CoA synthetase medium-chain family member 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001324371.1 | Intron | NP_001311300.1 | ||||
NM_001324372.1 | Intron | NP_001311301.1 | ||||
NM_001324373.1 | Intron | NP_001311302.1 | ||||
NM_017888.2 | Intron | NP_060358.2 | ||||
XM_006721056.3 | Intron | XP_006721119.1 |