Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGTTATTTCTGCCCCATCTGTGAA[A/G]CCTTGTCTGCCAATAGTGCCAAGTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300415 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MTM1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MTM1 - myotubularin 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000252.2 | Intron | NP_000243.1 | ||||
XM_005274687.2 | Intron | XP_005274744.1 | ||||
XM_011531171.1 | Intron | XP_011529473.1 | ||||
XM_011531172.1 | Intron | XP_011529474.1 | ||||
XM_011531173.2 | Intron | XP_011529475.1 | ||||
XM_017029547.1 | Intron | XP_016885036.1 | ||||
XM_017029548.1 | Intron | XP_016885037.1 | ||||
XM_017029549.1 | Intron | XP_016885038.1 | ||||
XM_017029550.1 | Intron | XP_016885039.1 | ||||
XM_017029551.1 | Intron | XP_016885040.1 |