Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAGATGATAACTCCTGACTTCTAG[A/G]CCCTATTTCTTCCAGAGCGCTTACT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 114025 MIM: 610867 | ||||||||||||||||||||
Literature Links: |
CTNNA2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CTNNA2 - catenin alpha 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001164883.1 | Intron | NP_001158355.1 | ||||
NM_001282597.2 | Intron | NP_001269526.1 | ||||
NM_001282598.1 | Intron | NP_001269527.1 | ||||
NM_001282599.1 | Intron | NP_001269528.1 | ||||
NM_001282600.1 | Intron | NP_001269529.1 | ||||
NM_001320810.1 | Intron | NP_001307739.1 | ||||
NM_004389.3 | Intron | NP_004380.2 | ||||
XM_011532555.2 | Intron | XP_011530857.1 | ||||
XM_011532556.2 | Intron | XP_011530858.1 | ||||
XM_011532557.2 | Intron | XP_011530859.1 | ||||
XM_017003403.1 | Intron | XP_016858892.1 | ||||
XM_017003404.1 | Intron | XP_016858893.1 | ||||
XM_017003405.1 | Intron | XP_016858894.1 | ||||
XM_017003406.1 | Intron | XP_016858895.1 |
LRRTM1 - leucine rich repeat transmembrane neuronal 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |