Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAACAGCATGTGAACGTGTGGAAA[A/G]AGTTAATGGGATAGTAAGTAGTTCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
C2orf16 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
C2orf16 - chromosome 2 open reading frame 16 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZNF512 - zinc finger protein 512 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001271286.1 | Intron | NP_001258215.1 | ||||
NM_001271287.1 | Intron | NP_001258216.1 | ||||
NM_001271288.1 | Intron | NP_001258217.1 | ||||
NM_001271289.1 | Intron | NP_001258218.1 | ||||
NM_001271318.1 | Intron | NP_001258247.1 | ||||
NM_032434.3 | Intron | NP_115810.2 |