Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTGTTCAACAGAGGCTTAAGGCCA[C/G]ATGTCCAAACTTGTCTCAATAAGGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611479 MIM: 612762 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GPN1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
GPN1 - GPN-loop GTPase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145047.1 | 4390 | Intron | NP_001138519.1 | |||
NM_001145048.1 | 4390 | Intron | NP_001138520.1 | |||
NM_001145049.1 | 4390 | Intron | NP_001138521.1 | |||
NM_007266.3 | 4390 | Intron | NP_009197.2 |
SUPT7L - SPT7-like STAGA complex gamma subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282729.1 | 4390 | UTR 3 | NP_001269658.1 | |||
NM_001282730.1 | 4390 | UTR 3 | NP_001269659.1 | |||
NM_001282731.1 | 4390 | UTR 3 | NP_001269660.1 | |||
NM_001282732.1 | 4390 | UTR 3 | NP_001269661.1 | |||
NM_014860.2 | 4390 | UTR 3 | NP_055675.1 | |||
XM_005264672.4 | 4390 | UTR 3 | XP_005264729.1 |
Set Membership: |
HapMap |