Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAATTTTTTTATGTTATTGTTTTC[A/G]GAGGAGATTTTCTGTGGGCAAATTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602691 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
NCOA1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
NCOA1 - nuclear receptor coactivator 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003743.4 | Intron | NP_003734.3 | ||||
NM_147223.2 | Intron | NP_671756.1 | ||||
NM_147233.2 | Intron | NP_671766.1 | ||||
XM_005264625.1 | Intron | XP_005264682.1 | ||||
XM_005264626.1 | Intron | XP_005264683.1 | ||||
XM_005264628.1 | Intron | XP_005264685.1 | ||||
XM_017005168.1 | Intron | XP_016860657.1 | ||||
XM_017005169.1 | Intron | XP_016860658.1 |
Set Membership: |
HapMap |