Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTCTGGCTCCTCAGAGCTTATCCT[A/G]CTTTCCCCCAGCCGTCCGGAGACGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ATP6V1C2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ATP6V1C2 - ATPase H+ transporting V1 subunit C2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001039362.1 | Intron | NP_001034451.1 | ||||
NM_144583.3 | Intron | NP_653184.2 | ||||
XM_011510339.2 | Intron | XP_011508641.1 | ||||
XM_011510340.2 | Intron | XP_011508642.1 | ||||
XM_011510341.2 | Intron | XP_011508643.1 | ||||
XM_017003745.1 | Intron | XP_016859234.1 |
Set Membership: |
HapMap |