Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGCTGGGGAGAAGGGCCATCCTGG[A/G]AGCCGGGCAGATGAGGACGGAACCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602635 MIM: 614988 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DEAF1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
DEAF1 - DEAF1, transcription factor | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
EPS8L2 - EPS8 like 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022772.3 | 188 | Intron | NP_073609.2 | |||
XM_017018130.1 | 188 | UTR 5 | XP_016873619.1 | |||
XM_017018131.1 | 188 | Intron | XP_016873620.1 | |||
XM_017018132.1 | 188 | Intron | XP_016873621.1 |
TMEM80 - transmembrane protein 80 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001042463.2 | 188 | Intron | NP_001035928.2 | |||
NM_001276253.1 | 188 | Intron | NP_001263182.1 | |||
NM_001276274.1 | 188 | Intron | NP_001263203.1 | |||
NM_174940.3 | 188 | Intron | NP_777600.3 | |||
XM_006718206.3 | 188 | Intron | XP_006718269.3 | |||
XM_017017600.1 | 188 | Intron | XP_016873089.1 |
Set Membership: |
HapMap |